2'-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)

Supplier: TCI America

B5674-1G B5674-200MG
TCB5674-1GEA 450.3 CAD
TCB5674-1G TCB5674-200MG
2'-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)
2'-Bromo-5'-phenyl-1,1':3',1''-terphenyl
Formula: C₂₄H₁₇Br
MDL Number: MFCD00092320
CAS Number: 10368-73-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 128.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR