9-Phenyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥96.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
P2571-1G
P2571-200MG
TCP2571-1GEA
455.04
CAD
TCP2571-1G
TCP2571-200MG
9-Phenyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥96.0% (by HPLC, total nitrogen)
9-Phenyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Formula:
C₃₀H₃₅B₂NO₄ |
CAS Number:
1035631-57-2 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (with Total Nitrogen) | min. 96.0 % |
Melting point | 235.0 to 240.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...