(3,5-Dibromophenyl)triphenylsilane ≥98.0% (by GC)
Supplier: TCI America
D5338-1G
D5338-200MG
76219-924EA
496.12
CAD
76219-924
76219-926
(3,5-Dibromophenyl)triphenylsilane ≥98.0% (by GC)
(3,5-Dibromophenyl)triphenylsilane
Formula:
C₂₄H₁₈Br₂Si |
CAS Number:
1030856-97-3 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 186.0 to 190.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...