Methyl (Z)-3-(p-Toluenesulfonyloxy)but-2-enoate ≥98.0% (by GC)
Supplier: TCI America
M3043-1G
M3043-5G
TCM3043-1GEA
161.16
CAD
TCM3043-1G
TCM3043-5G
Methyl (Z)-3-(p-Toluenesulfonyloxy)but-2-enoate ≥98.0% (by GC)
Methyl (Z)-3-(p-Toluenesulfonyloxy)but-2-enoate
Formula:
C₁₂H₁₄O₅S |
MDL Number:
MFCD30749294 CAS Number: 1029612-18-7 |
Specification Test Results
Purity (GC) | min. 98.0 % |
Melting point | 63.0 to 67.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...