9-(4-Biphenylyl)-3-(4-bromophenyl)carbazole ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
Synonyms:
9-([1,1'-Biphenyl]-4-yl)-3-(4-bromophenyl)-9H-carbazole
B5111-1G
B5111-200MG
TCB5111-1GEA
387.1
CAD
TCB5111-1G
TCB5111-200MG
9-(4-Biphenylyl)-3-(4-bromophenyl)carbazole ≥98.0% (by HPLC, total nitrogen)
9-(4-Biphenylyl)-3-(4-bromophenyl)carbazole
Formula:
C₃₀H₂₀BrN MW: 474.40 g/mol |
MDL Number:
MFCD22571689 CAS Number: 1028648-25-0 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 192.0 to 196.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...