11,12-Dihydro-11-phenylindolo[2,3-a]carbazole ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

D4860-1G D4860-200MG
TCD4860-1GEA 484.27 CAD
TCD4860-1G TCD4860-200MG
11,12-Dihydro-11-phenylindolo[2,3-a]carbazole ≥97.0% (by HPLC, total nitrogen)
11,12-Dihydro-11-phenylindolo[2,3-a]carbazole
Formula: C₂₄H₁₆N₂
MDL Number: MFCD22581305
CAS Number: 1024598-06-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR