4-Bromo-4'-[di(p-tolyl)amino]stilbene ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

B4906-1G B4906-200MG
TCB4906-1GEA 439.71 CAD
TCB4906-1G TCB4906-200MG
4-Bromo-4'-[di(p-tolyl)amino]stilbene ≥97.0% (by HPLC, total nitrogen)
4-Bromo-4'-[di(p-tolyl)amino]stilbene
Formula: C₂₈H₂₄BrN
MDL Number: MFCD28975103
CAS Number: 101186-77-0

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 155.0 to 159.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR