4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-(4-Pyridyl)phenylboronic acid Pinacol ester, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyridine, 4,4,5,5-Tetramethyl-2-[4-(4-pyridyl)phenyl]-1,3,2-dioxaborolane

T3400-1G T3400-200MG
TCT3400-1GEA 492.96 CAD
TCT3400-1G TCT3400-200MG
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine ≥98.0% (by GC, titration analysis)
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine
Formula: C₁₇H₂₀BNO₂
MW: 281.16 g/mol
MDL Number: MFCD12828171
CAS Number: 1009033-87-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR