Ethyl 7-Chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 7-Chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid Ethyl ester, Ethyl 7-chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate

E1180-25G E1180-5G
TCE1180-25GEA 201.61 CAD
TCE1180-25G TCE1180-5G
Ethyl 7-Chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate ≥98.0% (by HPLC, total nitrogen)
Ethyl 7-Chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate
Formula: C₁₇H₁₀ClF₃N₂O₃
MW: 382.72 g/mol
MDL Number: MFCD01863285
CAS Number: 100491-29-0
EINECS: 422-360-1
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 213.0 to 217.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR