2,2-Dimethyl-1,3-dioxacyclohept-5-ene ≥98.0% (by GC)

Supplier: TCI America
Warning

D5199-1G D5199-5G
76219-234EA 44.24 CAD
76219-234 76219-236
2,2-Dimethyl-1,3-dioxacyclohept-5-ene ≥98.0% (by GC)
2,2-Dimethyl-1,3-dioxacyclohept-5-ene
Formula: C₇H₁₂O₂
Boiling Pt: 147°C
Storage Temperature: Refrigerator
MDL Number: MFCD22054122
CAS Number: 1003-83-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR