2,8-Dibromodibenzofuran ≥98.0% (by GC)

Supplier: TCI America
Warning

D4821-1G D4821-5G
TCD4821-1GEA 83.74 CAD
TCD4821-1G TCD4821-5G
2,8-Dibromodibenzofuran ≥98.0% (by GC)
2,8-Dibromodibenzofuran
Formula: C₁₂H₆Br₂O
MDL Number: MFCD00093683
CAS Number: 10016-52-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 193.0 to 197.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR