2,2'-(Methylimino)bis(N,N-di-n-octylacetamide) ≥95.0% (by GC, titration analysis)

Supplier: TCI America

M2476-1ML M2476-5ML
TCM2476-1MLEA 281.24 CAD
TCM2476-1ML TCM2476-5ML
2,2'-(Methylimino)bis(N,N-di-n-octylacetamide) ≥95.0% (by GC, titration analysis)
2,2'-(Methylimino)bis(N,N-di-n-octylacetamide)
Formula: C₃₇H₇₅N₃O₂
MDL Number: MFCD28386110
CAS Number: 1000668-90-5

Order Now

Specification Test Results

Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR