9,10-Dihydro-9,9-dimethylacridine ≥98.0% (by GC)

Supplier: TCI America

D5028-1G D5028-200MG D5028-5G
TCD5028-1GEA 314.1 CAD
TCD5028-1G TCD5028-200MG TCD5028-5G
9,10-Dihydro-9,9-dimethylacridine ≥98.0% (by GC)
9,10-Dihydro-9,9-dimethylacridine
Formula: C₁₅H₁₅N
Boiling Pt: 155°C
MDL Number: MFCD00030130
CAS Number: 6267-02-3
EINECS: 228-436-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 125.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR