Niceritrol ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Pentaerythritol tetranicotinate

N0974-100MG N0974-25MG
TCN0974-100MGEA 685.72 CAD
TCN0974-100MG TCN0974-25MG
Niceritrol ≥98.0% (by HPLC)
Niceritrol
Formula: C₂₉H₂₄N₄O₈
MDL Number: MFCD00215982
CAS Number: 5868-05-3
EINECS: 227-519-7
Merck Index: 14,06497

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 9.50 to 10.50 %
Melting point 162.0 to 166.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR