4H-Indeno[1,2-b]thiophen-4-one ≥98.0% (by GC)

Supplier: TCI America

I1003-1G I1003-200MG
TCI1003-1GEA 298.62 CAD
TCI1003-1G TCI1003-200MG
4H-Indeno[1,2-b]thiophen-4-one ≥98.0% (by GC)
4H-Indeno[1,2-b]thiophen-4-one
Formula: C₁₁H₆OS
MW: 186.23 g/mol
CAS Number: 5706-08-1

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 99.0 to 103.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR