1,3-Benzenedisulfonamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B4917-1G B4917-200MG
76219-444EA 883.22 CAD
76219-444 76219-446
1,3-Benzenedisulfonamide ≥98.0% (by HPLC, total nitrogen)
1,3-Benzenedisulfonamide
Formula: C₆H₈N₂O₄S₂
MDL Number: MFCD00025383
CAS Number: 3701-01-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 233.0 to 237.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR