O-Benzyl-N,N'-diisopropylisourea ≥97.0% (by GC, titration analysis)
Supplier: TCI America
B4480-1ML
B4480-5ML
TCB4480-1MLEA
99.54
CAD
TCB4480-1ML
TCB4480-5ML
O-Benzyl-N,N'-diisopropylisourea ≥97.0% (by GC, titration analysis)
O-Benzyl-N,N'-diisopropylisourea
Formula:
C₁₄H₂₂N₂O Boiling Pt: 105°C |
MDL Number:
MFCD28386087 CAS Number: 2978-10-1 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...