Benfotiamine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B4711-25G B4711-5G
TCB4711-25GEA 350.05 CAD
TCB4711-25G TCB4711-5G
Benfotiamine ≥98.0% (by HPLC, titration analysis)
Benfotiamine
Formula: C₁₉H₂₃N₄O₆PS
Storage Temperature: Refrigerator
MDL Number: MFCD00057343
CAS Number: 22457-89-2
EINECS: 245-013-4
Merck Index: 14,01041

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR