Dexmedetomidine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

D5062-100MG D5062-20MG
TCD5062-100MGEA 327.06 CAD
TCD5062-100MG TCD5062-20MG
Dexmedetomidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Dexmedetomidine hydrochloride
Formula: C₁₃H₁₆N₂·HCl
Storage Temperature: Freezer
MDL Number: MFCD12912677
CAS Number: 145108-58-3
Merck Index: 14,02946

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 156.0 to 160.0 deg-C
Specific rotation [a]20/D +49.0 to +54.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR