Tetrandine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

T3321-1G T3321-200MG
TCT3321-1GEA 295.93 CAD
TCT3321-1G TCT3321-200MG
Tetrandine ≥98.0% (by HPLC, titration analysis)
Tetrandine
Formula: C₃₈H₄₂N₂O₆
MDL Number: MFCD08689909
CAS Number: 518-34-3
Merck Index: 14,09231

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 221.0 to 225.0 deg-C
Specific rotation [a]20/D +283 to +287 deg (C=1,CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR