2-Hydroxyflutamide ≥98.0% (by HPLC)

Supplier: TCI America
Warning

H1600-100MG H1600-25MG
TCH1600-100MGEA 1181.84 CAD
TCH1600-100MG TCH1600-25MG
2-Hydroxyflutamide ≥98.0% (by HPLC)
2-Hydroxyflutamide
Formula: C₁₁H₁₁F₃N₂O₄
MDL Number: MFCD00563126
CAS Number: 52806-53-8
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 9.00 to 10.00 %
Melting point 150.0 to 154.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR