(3R)-3-Aminobutan-1-ol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

A2951-1G A2951-5G
TCA2951-1GEA 153.73 CAD
TCA2951-1G TCA2951-5G
(3R)-3-Aminobutan-1-ol ≥98.0% (by GC, titration analysis)
(3R)-3-Aminobutan-1-ol
Formula: C₄H₁₁NO
MDL Number: MFCD13184351
CAS Number: 61477-40-5
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -12.0 to -10.0 deg (C=1,EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR