Moc-Val-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M2857-25G M2857-5G
TCM2857-25GEA 90.06 CAD
TCM2857-25G TCM2857-5G
Moc-Val-OH ≥98.0% (by HPLC, titration analysis)
Moc-Val-OH
Formula: C₇H₁₃NO₄
MDL Number: MFCD12795357
CAS Number: 74761-42-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 109.0 to 113.0 deg-C
Specific rotation [a]20/D -5.0 to -8.0 deg (C=1,AcOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR