Dimethylammonium dichlorotri(mu-chloro)bis{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}diruthenate(II)

Supplier: TCI America

N1024-1G N1024-200MG
TCN1024-1GEA 332.27 CAD
TCN1024-1G TCN1024-200MG
Dimethylammonium dichlorotri(mu-chloro)bis{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}diruthenate(II)
Formula: C₉₄H₉₆Cl₅NO₈P₄Ru₂
MDL Number: MFCD09753038
CAS Number: 944451-14-3

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR