Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]diruthenate(II)

Supplier: TCI America
Danger

N1022-1G N1022-200MG
TCN1022-1GEA 267.81 CAD
TCN1022-1G TCN1022-200MG
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]diruthenate(II)
Formula: C₇₈H₆₄Cl₅NO₈P₄Ru₂
MDL Number: MFCD09753037
CAS Number: 488809-34-3

Order Now

Specification Test Results

Appearance Orange to Brown powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR