Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)
Supplier: TCI America
|
Warning
|
Synonyms:
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II), [NH2Me2][{RuCl((R)-binap)}2(mu-Cl)3], [NH2Me2][(RuCl((R)-binap))2(mu-Cl)3]
N1015-1G
N1015-200MG
TCN1015-1GEA
231.47
CAD
TCN1015-1G
TCN1015-200MG
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)
Formula:
C₉₀H₇₂Cl₅NP₄Ru₂ |
MDL Number:
MFCD09753033 CAS Number: 199684-47-4 |
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to crystal |
IR | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...