Diacetato[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)

Supplier: TCI America

R0169-1G R0169-200MG
TCR0169-1GEA 213.3 CAD
TCR0169-1G TCR0169-200MG
Diacetato[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)
Diacetato[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)
MDL Number: MFCD09753021
CAS Number: 106681-15-6

Order Now

Specification Test Results

Appearance Dark green to Dark red to Black powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR