Diacetato[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)

Supplier: TCI America

R0168-1G R0168-200MG
TCR0168-1GEA 210.14 CAD
TCR0168-1G TCR0168-200MG
Diacetato[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)
Diacetato[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)
MDL Number: MFCD09753021
CAS Number: 116128-29-1

Order Now

Specification Test Results

Appearance Dark green to Dark red to Black powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR