Diacetato[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)

Supplier: TCI America

Synonyms: Ru(OAc)2[(R)-binap]

R0166-1G R0166-200MG
TCR0166-1GEA 235.42 CAD
TCR0166-1G TCR0166-200MG
Diacetato[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)
Diacetato[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)
Formula: C₄₈H₃₈O₄P₂Ru
MW: 841.83 g/mol
MDL Number: MFCD09753020
CAS Number: 325146-81-4

Order Now

Specification Test Results

Appearance Yellow to Amber to Dark purple powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR