Di-1-adamantylphosphine ≥95.0% (by GC)

Supplier: TCI America
Warning

D4781-1G D4781-200MG
TCD4781-1GEA 200.66 CAD
TCD4781-1G TCD4781-200MG
Di-1-adamantylphosphine ≥95.0% (by GC)
Di-1-adamantylphosphine
MDL Number: MFCD06658117
CAS Number: 131211-27-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 176.0 to 180.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR