2,3-Bis(2,6-di-i-propylphenylimino)butane ≥98.0% (by HPLC)
Supplier: TCI America
B5606-1G
B5606-200MG
76218-858EA
810.54
CAD
76218-858
76218-860
2,3-Bis(2,6-di-i-propylphenylimino)butane ≥98.0% (by HPLC)
2,3-Bis(2,6-di-i-propylphenylimino)butane
Formula:
C₂₈H₄₀N₂ |
MDL Number:
MFCD01862435 CAS Number: 74663-77-7 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 96.0 % |
Melting point | 99.0 to 103.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...