(R)-(+)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol ≥98.0% (by GC)

Supplier: TCI America
Warning

D4418-1G D4418-5G
TCD4418-1GEA 289.14 CAD
TCD4418-1G TCD4418-5G
(R)-(+)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol ≥98.0% (by GC)
(R)-(+)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol
Formula: C₂₀H₂₀Br₂O₂
Melting Pt: 149°C
MDL Number: MFCD03093977
CAS Number: 65355-08-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR