(R)-(-)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: (R)-N,N-Dimethyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine, (R)-MONOPHOS

D4992-1G D4992-5G
76219-100EA 90.06 CAD
76219-100 76219-102
(R)-(-)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine ≥98.0% (by HPLC, total nitrogen)
(R)-(-)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine
Formula: C₂₂H₁₈NO₂P
MW: 359.3576 g/mol
MDL Number: MFCD03426988
CAS Number: 157488-65-8

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -560 to -600 deg (C=0.3,CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR