Trihydroxyethylrutin ≥90.0% (by HPLC)
Supplier: TCI America
T3541-25G
T3541-5G
TCT3541-25GEA
262.28
CAD
TCT3541-25G
TCT3541-5G
Trihydroxyethylrutin ≥90.0% (by HPLC)
Trihydroxyethylrutin
Formula:
C₃₃H₄₂O₁₉ Melting Pt: 186°C Storage Temperature: Freezer |
MDL Number:
MFCD00893813 CAS Number: 7085-55-4 EINECS: 230-389-4 Merck Index: 14,09789 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 90.0 area% |
Specific rotation [a]20/D | -49.0 to -53.0 deg (C=1,Pyridine) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...