Dorsomorphin ≥97.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 6-(4-(2-(Piperidin-1-yl)ethoxy)phenyl)-3-(pyridin-4-yl)pyrazolo[1,5-a]pyrimidine, 6-[4-(2-Piperidin-1-ylethoxy)phenyl]-3-pyridin-4-ylpyrazolo[1,5-a]pyrimidine

D5394-10MG D5394-50MG
TCD5394-10MGEA 337.77 CAD
TCD5394-10MG TCD5394-50MG
Dorsomorphin ≥97.0% (by HPLC)
Dorsomorphin
Formula: C₂₄H₂₅N₅O
Storage Temperature: Refrigerator
MDL Number: MFCD08705402
CAS Number: 866405-64-3

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 16.50 to 18.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR