Genipin ≥97.0% (by GC)

Supplier: TCI America
Danger

Synonyms: Methyl (1S,2R,6S)-2-Hydroxy-9-(hydroxymethyl)-3-oxabicyclo[4.3.0]nona-4,8-diene-5-carboxylate, (1R,4aS,7aS)-Methyl 1-hydroxy-7-(hydroxymethyl)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylate

G0458-100MG G0458-25MG
TCG0458-100MGEA 287.56 CAD
TCG0458-100MG TCG0458-25MG
Genipin ≥97.0% (by GC)
Genipin
Formula: C₁₁H₁₄O₅
MDL Number: MFCD00888600
CAS Number: 6902-77-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 118.0 to 123.0 deg-C
Specific rotation [a]20/D +134 to +142 deg (C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR