(-)-Arctiin ≥97.0% (by HPLC)

Supplier: TCI America

A3118-10MG A3118-50MG
76219-294EA 74.26 CAD
76219-294 76219-296
(-)-Arctiin ≥97.0% (by HPLC)
(-)-Arctiin
Formula: C₂₇H₃₄O₁₁
Storage Temperature: Freezer
MDL Number: MFCD00870630
CAS Number: 20362-31-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 107.0 to 111.0 deg-C
Specific rotation [a]20/D -50.0 to -55.0 deg (C=2, MEOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR