Kenpaullone ≥97.0% (by HPLC)
Supplier: TCI America
|
Warning
|
Synonyms:
9-Bromo-7,12-dihydroindolo[3,2-d][1]benzazepin-6(5H)-one, 9-Bromo-7,12-dihydrobenzo[2,3]azepino[4,5-b]indol-6(5H)-one
K0052-10MG
K0052-50MG
TCK0052-10MGEA
239.69
CAD
TCK0052-10MG
TCK0052-50MG
Kenpaullone ≥97.0% (by HPLC)
Kenpaullone
Formula:
C₁₆H₁₁BrN₂O MW: 327.18 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD02683595 CAS Number: 142273-20-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Elemental analysis (Nitrogen) | 8.00 to 9.00 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...