Spectinomycin dihydrochloride pentahydrate ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

S0584-25G S0584-5G
TCS0584-25GEA 440.82 CAD
TCS0584-25G TCS0584-5G
Spectinomycin dihydrochloride pentahydrate ≥98.0% (by total nitrogen and titration analysis)
Spectinomycin dihydrochloride pentahydrate
Formula: C₁₄H₂₄N₂O₇·2HCl·5H₂O
Melting Pt: 207°C
Storage Temperature: Refrigerator
MDL Number: MFCD00150886
CAS Number: 22189-32-8
Merck Index: 14,08739

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +15.0 to +21.0 deg (C=1,H2O)
Water 16.0 to 20.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR