Sisomicin sulphate Salt ≥98.0% (by HPLC)

Supplier: TCI America
Danger

I1049-1G I1049-250MG
TCI1049-1GEA 231.44 CAD
TCI1049-1G TCI1049-250MG
Sisomicin sulphate Salt ≥98.0% (by HPLC)
Sisomicin sulphate Salt
Formula: (C₁₉H₃₇N₅O₇)₂·5H₂SO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00058308
CAS Number: 53179-09-2
EINECS: 258-414-4
Merck Index: 14,08550

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +98.0 to +104.0 deg (C=1, H2O)
Drying loss max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR