Roxithromycin ≥95.0% (by HPLC)

Supplier: TCI America
Warning

R0164-25G R0164-5G
TCR0164-25GEA 366.56 CAD
TCR0164-25G TCR0164-5G
Roxithromycin ≥95.0% (by HPLC)
Roxithromycin
Formula: C₄₁H₇₆N₂O₁₅
Melting Pt: 120°C
Storage Temperature: Refrigerator
MDL Number: MFCD00214389
CAS Number: 80214-83-1
Merck Index: 14,08276

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 3.10 to 3.60 %
Specific rotation [a]20/D -93.0 to -96.0 deg (C=1, Acetone) (calcd.on anh.substance)
Water max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR