3-(4-Pyridyl)-L-alanine dihydrochloride ≥96.0% (by HPLC, total nitrogen)
Supplier: TCI America
P2136-1G
P2136-5G
TCP2136-1GEA
221.2
CAD
TCP2136-1G
TCP2136-5G
3-(4-Pyridyl)-L-alanine dihydrochloride ≥96.0% (by HPLC, total nitrogen)
3-(4-Pyridyl)-L-alanine dihydrochloride
Formula:
C₈H₁₀N₂O₂·2HCl |
MDL Number:
MFCD13184932 CAS Number: 178933-04-5 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (with Total Nitrogen) | min. 96.0 % |
Specific rotation [a]20/D | +23.0 to +26.0 deg (C=2,H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...