Polyethylene glycol dimethylether, M.W. ∼ 240
Supplier: TCI America
P2241-25G
P2241-500G
TCP2241-25GEA
48.98
CAD
TCP2241-25G
TCP2241-500G
Polyethylene glycol dimethylether, M.W. ∼ 240
Polyethylene glycol dimethylether 150
Formula:
H₃C(OCH₂CH₂)nOCH₃ Flash Pt: 156°C |
MDL Number:
MFCD00147972 CAS Number: 24991-55-7 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Average molecular weight | 230 to 250 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...