Procarbazine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
N-Isopropyl-4-[(2-methylhydrazino)methyl]benzamide, N-Isopropyl-4-((2-methylhydrazinyl)methyl)benzamide hydrochloride, Procarbazine HCl
P2550-100MG
P2550-500MG
TCP2550-100MGEA
66.76
CAD
TCP2550-100MG
TCP2550-500MG
Procarbazine hydrochloride ≥98.0% (by HPLC, titration analysis)
Procarbazine hydrochloride
Formula:
C₁₂H₁₉N₃O·HCl MW: 257.76 g/mol Melting Pt: 226°C |
MDL Number:
MFCD00072082 CAS Number: 366-70-1 EINECS: 206-678-6 Merck Index: 14,07758 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...