(S)-3-Methylmorpholine hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
M2848-1G
M2848-5G
76220-280EA
210.14
CAD
76220-280
76220-282
(S)-3-Methylmorpholine hydrochloride ≥98.0% (by titrimetric analysis)
(S)-3-Methylmorpholine hydrochloride
Formula:
C₅H₁₁NO·ClH Storage Temperature: Refrigerator |
MDL Number:
MFCD18382512 CAS Number: 1022094-03-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 125.0 to 130.0 deg-C |
Specific rotation [a]20/D | -2.0 to -5.0 deg (C=1, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...