1,4-Diazabicyclo[2.2.2]octanebis(sulfur dioxide)adduct adduct ≥94.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B3960-1G B3960-5G
TCB3960-1GEA 67.94 CAD
TCB3960-1G TCB3960-5G
1,4-Diazabicyclo[2.2.2]octanebis(sulfur dioxide)adduct adduct ≥94.0% (by titrimetric analysis)
1,4-Diazabicyclo[2.2.2]octanebis(sulfur dioxide)adduct
Formula: C₆H₁₂N₂O₄S₂
Storage Temperature: Refrigerator
MDL Number: MFCD22377802
CAS Number: 119752-83-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Iodometric back titration) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR