Lilolidine ≥98.0% (by GC)

Supplier: TCI America
Warning

L0274-1G L0274-5G
TCL0274-1GEA 127.35 CAD
TCL0274-1G TCL0274-5G
Lilolidine ≥98.0% (by GC)
Lilolidine
Formula: C₁₁H₁₁N
MDL Number: MFCD09841910
CAS Number: 5840-01-7

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 84.0 to 88.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR