2-Cyclohepten-1-one ≥85.0% (by GC)

Supplier: TCI America

C3180-1G C3180-5G
TCC3180-1GEA 137.46 CAD
TCC3180-1G TCC3180-5G
2-Cyclohepten-1-one ≥85.0% (by GC)
2-Cyclohepten-1-one
Formula: C₇H₁₀O
Boiling Pt: 183°C
Flash Pt: 68°C
Storage Temperature: Refrigerator
MDL Number: MFCD00004157
CAS Number: 1121-66-0
EINECS: 214-334-1

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR