Sodium bis(trifluoromethylsulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

S0989-1G S0989-5G
TCS0989-1GEA 135.88 CAD
TCS0989-1G TCS0989-5G
Sodium bis(trifluoromethylsulfonyl)imide ≥98.0% (by titrimetric analysis)
Sodium bis(trifluoromethylsulfonyl)imide
Formula: C₂F₆NNaO₄S₂
Melting Pt: 257°C
MDL Number: MFCD14156021
CAS Number: 91742-21-1
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR