Calcium bis(trifluoromethylsulfonyl)imide ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

C3263-1G C3263-5G
TCC3263-1GEA 173.8 CAD
TCC3263-1G TCC3263-5G
Calcium bis(trifluoromethylsulfonyl)imide ≥97.0% (by titrimetric analysis)
Calcium bis(trifluoromethylsulfonyl)imide
Formula: C₄CaF₁₂N₂O₈S₄
MDL Number: MFCD14156019
CAS Number: 165324-09-4
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 97.0 %
Purity (Ion exchange titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR